4-(Aminomethyl)piperidine 96%

Supplier: Ambeed
Danger

A156621-1G A156621-25G A156621-5G
77286-296EA 6 USD
77286-296 77286-298 77286-300
4-(Aminomethyl)piperidine 96%
4-(Aminomethyl)piperidine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MDL Number: MFCD00006007
CAS Number: 7144-05-0
UN: 3259
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR