2-Bromo-5-(trifluoromethoxy)aniline 98%
Supplier: Ambeed
|
Warning
|
A156376-100G
A156376-10G
A156376-1G
A156376-25G
A156376-5G
77259-472EA
95.79
USD
77259-472
77259-474
77259-476
77259-478
77259-480
2-Bromo-5-(trifluoromethoxy)aniline 98%
2-Bromo-5-(trifluoromethoxy)aniline
Precaución: for Research Use Only
Formula:
C₇H₅BrF₃NO |
MDL Number:
MFCD08458013 CAS Number: 887267-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...