2-Methyl-5-thiazolecarboxylic acid 98%

Supplier: Ambeed
Warning

A156337-100G A156337-10G A156337-1G A156337-25G A156337-5G
77259-462EA 453.2 USD
77259-462 77259-464 77259-466 77259-468 77259-470
2-Methyl-5-thiazolecarboxylic acid 98%
2-Methyl-5-thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₂S
MDL Number: MFCD01760300
CAS Number: 40004-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR