2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone 95%

Supplier: Ambeed
Warning

A156294-100G A156294-10G A156294-1G A156294-25G A156294-5G
77247-474EA 348.14 USD
77247-474 77247-476 77247-478 77247-480 77247-482
2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone 95%
2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone

Precaución: for Research Use Only

Formula: C₃₄H₃₄O₆
MW: 538.6302 g/mol
MDL Number: MFCD08703228
CAS Number: 13096-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR