2,4-Dimethoxyphenol 97%

Supplier: Ambeed
Warning

A156122-10G A156122-1G A156122-250MG A156122-25G A156122-5G
77247-464EA 68 USD
77247-464 77247-466 77247-468 77247-470 77247-472
2,4-Dimethoxyphenol 97%
2,4-Dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MDL Number: MFCD00060637
CAS Number: 13330-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR