3-(Trifluoromethyl)benzamide 98%

Supplier: Ambeed
Warning

A156067-100G A156067-1G A156067-1MG A156067-25G A156067-5G
77235-662EA 99 USD
77235-662 77235-664 77235-666 77235-668 77235-670
3-(Trifluoromethyl)benzamide 98%
3-(Trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MDL Number: MFCD00014802
CAS Number: 1801-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR