2-Bromo-4-(trifluoromethyl)phenylacetonitrile 98%

Supplier: Ambeed
Warning

A156061-1G A156061-250MG
77259-452EA 113.3 USD
77259-452 77259-454
2-Bromo-4-(trifluoromethyl)phenylacetonitrile 98%
2-Bromo-4-(trifluoromethyl)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₅BrF₃N
MW: 264.04 g/mol
MDL Number: MFCD03094308
CAS Number: 474024-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR