2-(1H-Imidazol-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A156049-10G A156049-1G A156049-250MG A156049-25G A156049-5G
77235-652EA 158.62 USD
77235-652 77235-654 77235-656 77235-658 77235-660
2-(1H-Imidazol-2-yl)pyridine 97%
2-(1H-Imidazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MW: 145.161 g/mol
MDL Number: MFCD05863221
CAS Number: 18653-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR