Elbasvir 98%

Supplier: Ambeed
Warning

Synonyms: MK-8742, Dimethyl ((2S,2'S)-((2S,2'S)-2,2'-(5,5'-((S)-6-phenyl-6H-benzo[5,6][1,3]oxazino[3,4-a]indole-3,10-diyl)bis(1H-imidazole-5,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methyl-1-oxobutane-2,1-diyl))dicarbamate

A155998-10MG A155998-1MG A155998-2MG A155998-5MG
77235-644EA 45.32 USD
77235-644 77235-646 77235-648 77235-650
Elbasvir 98%
Elbasvir

Precaución: for Research Use Only

Formula: C₄₉H₅₅N₉O₇
MW: 882.02 g/mol
MDL Number: MFCD30829850
CAS Number: 1370468-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR