(R)-Methyl 2-((tert-butyldimethylsilyl)oxy)propanoate 97%

Supplier: Ambeed
Warning

A155826-100MG A155826-1G A155826-250MG
77259-440EA 33.99 USD
77259-440 77259-442 77259-444
(R)-Methyl 2-((tert-butyldimethylsilyl)oxy)propanoate 97%
(R)-Methyl 2-((tert-butyldimethylsilyl)oxy)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₃Si
MW: 218.37 g/mol
MDL Number: MFCD28402584
CAS Number: 171230-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR