trans-4-(4-n-Propylcyclohexyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(trans-4-Propylcyclohexyl)benzoic acid, 4-(trans-4-n-Propylcyclohexyl)benzoic acid

A155805-10G A155805-1G A155805-25G A155805-5G
77273-130EA 8 USD
77273-130 77273-132 77273-134 77273-136
trans-4-(4-n-Propylcyclohexyl)benzoic acid 98%
trans-4-(4-n-Propylcyclohexyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₂
MW: 246.35 g/mol
MDL Number: MFCD06658177
CAS Number: 65355-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR