1,3-Dichloro-2-fluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A155741-100G A155741-10G A155741-1G A155741-25G A155741-500G A155741-5G
77286-254EA 41 USD
77286-254 77286-256 77286-258 77286-260 77286-262 77286-264
1,3-Dichloro-2-fluoro-5-nitrobenzene 98%
1,3-Dichloro-2-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FNO₂
MDL Number: MFCD00129963
CAS Number: 3107-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR