Vipadenant 98%

Supplier: Ambeed
Warning

Synonyms: BIIB014, CEB-4520, BIIB-014, 3-[(4-Amino-3-methylphenyl)methyl]-7-(2-furanyl)-3H-1,2,3-triazolo[4,5-d]pyrimidin-5-amine, 3-(4-Amino-3-methylbenzyl)-7-(furan-2-yl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-5-amine

A155723-10MG A155723-25MG A155723-50MG A155723-5MG A155723-1MG A155723-2MG A155723-250MG
77235-628EA 113.3 USD
77235-628 77235-630 77235-632 77235-634 77615-270 77615-272 77746-072
Vipadenant 98%
Vipadenant

Precaución: for Research Use Only

Formula: C₁₆H₁₅N₇O
MW: 321.3 g/mol
MDL Number: MFCD12024697
CAS Number: 442908-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR