2-(4-Bromophenyl)-1-phenylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromophenyl)-1-phenyl-1H-benzoimidazole

A155525-10G A155525-1G A155525-25G A155525-5G
77235-616EA 61.8 USD
77235-616 77235-618 77235-620 77235-622
2-(4-Bromophenyl)-1-phenylbenzimidazole 98%
2-(4-Bromophenyl)-1-phenylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₉H₁₃BrN₂
MW: 349.23 g/mol
MDL Number: MFCD09261342
CAS Number: 2620-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR