2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile 98%

Supplier: Ambeed
Danger

A155444-250MG A155444-5G A155444-1G
77247-446EA 37.08 USD
77247-446 77247-448 77746-068
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile 98%
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂S
MDL Number: MFCD00121495
CAS Number: 70291-62-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR