2-Fluoroethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-Aminoethyl Fluoride hydrochloride, 2-Fluoroethanamine hydrochloride, 2-Fluoroethylammonium hydrochloride

A155303-10G A155303-1G A155303-250MG A155303-25G A155303-5G
77259-426EA 96.82 USD
77259-426 77259-428 77259-430 77259-432 77259-434
2-Fluoroethylamine hydrochloride 95%
2-Fluoroethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₂H₆FN·HCl
MW: 99.53 g/mol
MDL Number: MFCD00008161
CAS Number: 460-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR