(S)-1-Methoxypropan-2-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A155257-100MG
A155257-1G
A155257-250MG
A155257-5G
77259-418EA
15.45
USD
77259-418
77259-420
77259-422
77259-424
(S)-1-Methoxypropan-2-amine hydrochloride 97%
(S)-1-Methoxypropan-2-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₂ClNO MW: 125.59 g/mol |
MDL Number:
MFCD20502233 CAS Number: 1162054-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...