6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-6-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A155192-100MG A155192-1G A155192-250MG
77247-438EA 29 USD
77247-438 77247-440 77247-442
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-6-carboxylic acid hydrochloride 97%
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-6-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O₂
MDL Number: MFCD30181971
CAS Number: 1965310-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR