XY1 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Naphthalen-2-yl)-3-(2-oxo-2-(pyrrolidin-1-yl)ethyl)urea

A155170-10MG A155170-1MG A155170-50MG A155170-5MG
77235-608EA 136.99 USD
77235-608 77235-610 77235-612 77235-614
XY1 98%
XY1

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₂
MW: 297.35 g/mol
MDL Number: MFCD28397755
CAS Number: 1624117-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR