(R)-3,3'-Bis(4-methoxyphenyl)-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A155163-100MG A155163-1G A155163-250MG
77247-432EA 49.44 USD
77247-432 77247-434 77247-436
(R)-3,3'-Bis(4-methoxyphenyl)-[1,1'-binaphthalene]-2,2'-diol 98%
(R)-3,3'-Bis(4-methoxyphenyl)-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₃₄H₂₆O₄
MW: 498.56 g/mol
MDL Number: MFCD30541640
CAS Number: 756491-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR