Benzo[d]thiazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A155082-250MG A155082-1G
77235-602EA 265 USD
77235-602 77746-066
Benzo[d]thiazole-4-carbaldehyde 97%
Benzo[d]thiazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅NOS
MDL Number: MFCD22394756
CAS Number: 1213833-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR