2-(3-Aminophenyl)-1H-benzo[d]imidazol-6-amine 95%

Supplier: Ambeed
Warning

A155013-1G A155013-5G
77235-598EA 37.08 USD
77235-598 77235-600
2-(3-Aminophenyl)-1H-benzo[d]imidazol-6-amine 95%
2-(3-Aminophenyl)-1H-benzo[d]imidazol-6-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₄
MW: 224.26 g/mol
MDL Number: MFCD13874743
CAS Number: 13676-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR