Triethylene glycol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol
A154997-1000G
A154997-100G
A154997-500G
77247-426EA
11.33
USD
77247-426
77247-428
77247-430
Triethylene glycol 98%
Triethylene glycol
Precaución: for Research Use Only
Formula:
C₆H₁₄O₄ MW: 150.17 g/mol |
MDL Number:
MFCD00002880 CAS Number: 112-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...