(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclobutylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Cyclobutyl-Gly-OH

A154884-100MG A154884-50MG A154884-1G A154884-5G A154884-250MG
77235-594EA 37 USD
77235-594 77235-596 77567-256 77567-258 77615-266
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclobutylacetic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclobutylacetic acid

Precaución: For research use only.

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD09839305
CAS Number: 1391630-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR