2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane 98%

Supplier: Ambeed
Danger

Synonyms: 3-Perfluorobutyl-1,2-epoxypropane

A154803-1G A154803-5G
77286-226EA 13.39 USD
77286-226 77286-228
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane 98%
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane

Precaución: for Research Use Only

Formula: C₇H₅F₉O
MW: 276.10 g/mol
MDL Number: MFCD00236085
CAS Number: 81190-28-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR