9-(3-Fluorophenyl)-9H-carbazole 97%

Supplier: Ambeed
Warning

A154796-100MG A154796-1G A154796-250MG
77247-416EA 12.36 USD
77247-416 77247-418 77247-420
9-(3-Fluorophenyl)-9H-carbazole 97%
9-(3-Fluorophenyl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂FN
MW: 261.29 g/mol
MDL Number: MFCD30543717
CAS Number: 81329-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR