2-Methyl-5-nitro-2H-indazole 95%

Supplier: Ambeed
Warning

A154787-1G A154787-250MG
77273-110EA 27 USD
77273-110 77273-112
2-Methyl-5-nitro-2H-indazole 95%
2-Methyl-5-nitro-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MDL Number: MFCD00051809
CAS Number: 5228-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR