3-Bromo-5-azaindole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-1H-pyrrolo[3,2-c]pyridine

A154725-10G A154725-250MG A154725-25G A154725-5G A154725-1G
77273-102EA 400.67 USD
77273-102 77273-104 77273-106 77273-108 77748-064
3-Bromo-5-azaindole 97%
3-Bromo-5-azaindole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.032 g/mol
MDL Number: MFCD08690126
CAS Number: 23612-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR