2',3',5'-triacetyl-5-Azacytidine 97%

Supplier: Ambeed
Warning

Synonyms: 2′,3′,5′-Triacetyl-5-azacytidine, (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(4-amino-2-oxo-1,3,5-triazin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate

A154661-250MG A154661-1G A154661-25G A154661-5G A154661-100MG
77235-584EA 79 USD
77235-584 77592-898 77615-262 77615-264 77746-061
2',3',5'-triacetyl-5-Azacytidine 97%
2',3',5'-triacetyl-5-Azacytidine

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₄O₈
MDL Number: MFCD11113159
CAS Number: 10302-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR