2,3-Difluoro-4-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A154583-1G
A154583-250MG
A154583-5G
77247-410EA
46.35
USD
77247-410
77247-412
77247-414
2,3-Difluoro-4-hydroxybenzaldehyde 97%
2,3-Difluoro-4-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄F₂O₂ |
MDL Number:
MFCD08235194 CAS Number: 676500-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...