2-Chloro-3-cyano-4-picoline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-methylpyridine-3-carbonitrile
A154556-1G
A154556-250MG
A154556-5G
77259-410EA
72.1
USD
77259-410
77259-412
77259-414
2-Chloro-3-cyano-4-picoline 97%
2-Chloro-3-cyano-4-picoline
Precaución: for Research Use Only
Formula:
C₇H₅ClN₂ MW: 152.5810 g/mol |
MDL Number:
MFCD07082888 CAS Number: 65169-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...