(E)-4-(Piperidin-1-yl)but-2-enoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A154451-100MG
A154451-1G
A154451-250MG
A154451-25G
A154451-5G
77235-568EA
5.15
USD
77235-568
77235-570
77235-572
77235-574
77235-576
(E)-4-(Piperidin-1-yl)but-2-enoic acid hydrochloride 97%
(E)-4-(Piperidin-1-yl)but-2-enoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₆ClNO₂ MW: 205.68 g/mol |
MDL Number:
MFCD14636476 CAS Number: 197892-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...