Methyl-4-aminobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-aminobenzoate

A154368-100G A154368-10G A154368-1G A154368-500G A154368-5G A154368-25G
77247-400EA 39.14 USD
77247-400 77247-402 77247-404 77247-406 77247-408 77318-526
Methyl-4-aminobenzoate 98%
Methyl-4-aminobenzoate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00007891
CAS Number: 619-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR