2-Amino-3,5-dimethylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethylanthranilic acid

A154344-1G A154344-5G
77273-086EA 12.36 USD
77273-086 77273-088
2-Amino-3,5-dimethylbenzoic acid 97%
2-Amino-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00017099
CAS Number: 14438-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR