2,5-Dithiobiurea 95%

Supplier: Ambeed
Warning

A154323-100G A154323-25G A154323-5G
77247-394EA 31 USD
77247-394 77247-396 77247-398
2,5-Dithiobiurea 95%
2,5-Dithiobiurea

Precaución: for Research Use Only

Formula: C₂H₆N₄S₂
MDL Number: MFCD00014471
CAS Number: 142-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR