2-Methylquinazolin-4-ol 95%

Supplier: Ambeed
Warning

A154228-10G A154228-1G A154228-250MG A154228-5G A154228-25G
77273-078EA 88.58 USD
77273-078 77273-080 77273-082 77273-084 77748-063
2-Methylquinazolin-4-ol 95%
2-Methylquinazolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.18 g/mol
MDL Number: MFCD05270928
CAS Number: 1769-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR