2,5-Dibromophenylboronic acid 97%

Supplier: Ambeed
Warning

A154115-10G A154115-250MG
77273-068EA 127.72 USD
77273-068 77273-070
2,5-Dibromophenylboronic acid 97%
2,5-Dibromophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBr₂O₂
MDL Number: MFCD09743927
CAS Number: 1008106-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR