2-Isobutyl-3-methylpyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-3-isobutylpyrazine

A154104-1G A154104-10G A154104-5G
77259-384EA 9.27 USD
77259-384 77748-061 77748-062
2-Isobutyl-3-methylpyrazine 97%
2-Isobutyl-3-methylpyrazine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MW: 150.2209 g/mol
MDL Number: MFCD00059770
CAS Number: 13925-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR