2-Iodo-4-methoxy-1-(methylsulfonyl)benzene 97%

Supplier: Ambeed
Warning

A154095-100MG A154095-1G A154095-250MG
77247-388EA 33.99 USD
77247-388 77247-390 77247-392
2-Iodo-4-methoxy-1-(methylsulfonyl)benzene 97%
2-Iodo-4-methoxy-1-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉IO₃S
MW: 312.125 g/mol
MDL Number: MFCD30181850
CAS Number: 1965309-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR