1-Methyl-1H-pyrazole-3-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A153883-100MG
A153883-1G
A153883-250MG
77235-560EA
21.63
USD
77235-560
77235-562
77235-564
1-Methyl-1H-pyrazole-3-carbonitrile 95%
1-Methyl-1H-pyrazole-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₅H₅N₃ |
MDL Number:
MFCD00466343 CAS Number: 79080-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...