Trans-4-cyano-3-fluorophenyl 4-ethylcyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A153830-10G A153830-25G A153830-5G
77286-210EA 21.63 USD
77286-210 77286-212 77286-214
Trans-4-cyano-3-fluorophenyl 4-ethylcyclohexanecarboxylate 97%
Trans-4-cyano-3-fluorophenyl 4-ethylcyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₈FNO₂
MW: 275.32 g/mol
MDL Number: MFCD11053368
CAS Number: 90525-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR