2-Amino-3-bromo-5-methylbenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A153718-100G
A153718-10G
A153718-1G
A153718-250MG
A153718-25G
A153718-500G
A153718-5G
77235-530EA
450.11
USD
77235-530
77235-532
77235-534
77235-536
77235-538
77235-540
77235-542
2-Amino-3-bromo-5-methylbenzoic acid 98%
2-Amino-3-bromo-5-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol |
MDL Number:
MFCD00051705 CAS Number: 13091-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...