2-Bromo-4-chlorobenzonitrile 98%

Supplier: Ambeed
Warning

A153607-10G A153607-1G A153607-25G A153607-5G A153607-100G A153607-500G
77259-352EA 11 USD
77259-352 77259-354 77259-356 77259-358 77746-056 77746-057
2-Bromo-4-chlorobenzonitrile 98%
2-Bromo-4-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrClN
MDL Number: MFCD00158966
CAS Number: 57381-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR