Fmoc-Tyr(Bzl)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-O-benzyl-L-tyrosine

A153606-100G A153606-10G A153606-1G A153606-5G A153606-25G
77286-198EA 191 USD
77286-198 77286-200 77286-202 77615-252 77748-055
Fmoc-Tyr(Bzl)-OH 98%
Fmoc-Tyr(Bzl)-OH

Precaución: for Research Use Only

Formula: C₃₁H₂₇NO₅
MDL Number: MFCD00065682
CAS Number: 71989-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR