4-Amino-3-bromopyridine 95%

Supplier: Ambeed
Warning

A153538-100G A153538-10G A153538-1G A153538-25G A153538-5G
77273-054EA 115 USD
77273-054 77273-056 77273-058 77273-060 77273-062
4-Amino-3-bromopyridine 95%
4-Amino-3-bromopyridine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MDL Number: MFCD02068297
CAS Number: 13534-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR