2-Bromo-6-(methylthio)pyridine 95%

Supplier: Ambeed
Warning

A153504-1G A153504-250MG A153504-5G
77259-346EA 60 USD
77259-346 77259-348 77259-350
2-Bromo-6-(methylthio)pyridine 95%
2-Bromo-6-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNS
MW: 204.0900 g/mol
MDL Number: MFCD09265530
CAS Number: 74134-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR