Methyl 2-(hydroxymethyl)isonicotinate 97%
Supplier: Ambeed
|
Warning
|
A153405-250MG
A153405-5G
A153405-1G
77259-338EA
108.15
USD
77259-338
77549-934
77748-054
Methyl 2-(hydroxymethyl)isonicotinate 97%
Methyl 2-(hydroxymethyl)isonicotinate
Precaución: for Research Use Only
Formula:
C₈H₉NO₃ MW: 167.16 g/mol |
MDL Number:
MFCD11848098 CAS Number: 58481-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...