Morpholine-2-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A153384-100MG
A153384-250MG
77273-046EA
12.36
USD
77273-046
77273-048
Morpholine-2-carboxylic acid hydrochloride 95%
Morpholine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₀ClNO₃ MW: 167.59 g/mol |
MDL Number:
MFCD06410629 CAS Number: 878010-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...