AG-120 (Ivosidenib) 98%

Supplier: Ambeed
Warning

A153326-100MG A153326-10MG A153326-1G A153326-250MG A153326-50MG A153326-5MG A153326-1MG
77235-514EA 72 USD
77235-514 77235-516 77235-518 77235-520 77235-522 77235-524 77746-054
AG-120 (Ivosidenib) 98%
AG-120 (Ivosidenib)

Precaución: for Research Use Only

Formula: C₂₈H₂₂ClF₃N₆O₆
MDL Number: MFCD29044889
CAS Number: 1448347-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR