AZD1152-HQPA 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-((7-(3-(Ethyl(2-hydroxyethyl)amino)propoxy)quinazolin-4-yl)amino)-1H-pyrazol-3-yl)-N-(3-fluorophenyl)acetamide, Barasertib-HQPA, Barasertib

A153304-10MG A153304-1MG A153304-5MG
77235-508EA 28.84 USD
77235-508 77235-510 77235-512
AZD1152-HQPA 98%
AZD1152-HQPA

Precaución: for Research Use Only

Formula: C₂₆H₃₀FN₇O₃
MW: 507.56 g/mol
MDL Number: MFCD17392583
CAS Number: 722544-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR